Learn More
Fisherbrand™ Cryogenic Labels for Cryogenic Storage
Sfoglia Tutto Fisherbrand Prodotti
Descrizione
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy to use dispenser box with 20 sheets per pack.
Ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. Suitable for use on microcentrifuge tubes, cryovials, racks, boxes and microtiter plates. Adhesive withstands temperatures from -196° to +121°C (-321° to +250°F)
- No need to self-laminate
- Thinner labels allow tubes to fit most storage systems
- Available in a full range of colors and sizes
- Guaranteed not to fall off under normal use (provided user instructions are followed)
- Also offered in laser printer-compatible sheets (see Cat. Nos. 15-930 and 15-940 Series)
Cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications
Specifica
Specifica
| Colore | Yellow |
| Formato | Label |
| Product Type | Cryogenic Label |
| Da utilizzare con (applicazione) | Cryostorage, Liquid Nitrogen, refrigeration and ultra-low temperature freezer applications |
| Da utilizzare con (apparecchiatura) | 1.5 to 2 mL tubes |
| Quantità | 1000/Roll |
| Dimensioni (sistema imperiale) | 0.5 x 1.25 in. |
| Dimensioni (sistema metrico) | 1.2 x 3.1 cm |
| Intervallo di temperatura (sistema imperiale) | -321 to 250°F |
| Temperature Range (Metric) | -196°C to +121°C |
| Vedi altri risultati |