Learn More
Fisherbrand™ Glass Beaker with Cover
The Fisherbrand™ Glass Beaker with Cover is an ideal accessory for the Fisherbrand™ 11211 ultrasonic cleaner. This is a safe and economical choice for any laboratory.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Clear glass for easy viewing
- Stable design prevents tipping
- Easy to open and close lid
- Graduated design
Specifica
Beaker | |
33.81 oz. | |
Lid |
1 L | |
FB11211 Ultrasonic Cleaning Unit | |
Glass |