Learn More
Motic™ FITC Filter Block, BA310E
For detection of FITC fluorophore. Motic™ FITC Filter Block, BA310E is suitable for BA310E Microscopes.
Marca: Motic™ 1101001902923
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Epi-LED fluorescence equipment for FITC (blue) excitation.
- Block houses an excitation filter, an emission filter, and a dichroic beam splitter.
- Suitable for BA-310E Microscopes.
Specifica
Filter Block, FITC | |
BA-310E Microscopes | |
Excitation Filter, Emission Filter, Dichroic Beam splitter |