Learn More
Thermo Scientific™ FlipStrip™ Microtube Racks with Lid
Sfoglia Tutto MBP Molecular BioProducts Prodotti
Click to view available options
Colore:
Blue
Fluorescent Green
Fluorescent Green, Fluorescent Orange, Fluorescent Pink, Purple
Fluorescent Orange
Fluorescent Pink
Giallo
Green
Purple
Red
Red, Green, Yellow, or Blue
Dimensione della confezione:
Pezzo
pezzi 12
Descrizione
Hold 0.2mL strips and 1.5mL tubes on one side of the Thermo Scientific™ FlipStrip Microtube Rack with Lid, and 0.5mL tubes on the other. The rack is made from 100% recycled plastic.
- Holds 0.2mL strips and 1.5mL tubes on one side, 0.5mL tubes on other
- Stable, low-profile design with an alphanumeric grid for easy tube identification
- Lid protects samples and allows racks to be stacked
- Several colors available
Specifica
Specifica
| Product Type | Flipstrip Rack |
| Colore | Fluorescent Green |
| Capacità | Microtubes (0.5 mL), Microtubes (1.5 mL), Microtubes (2 mL) |
| Materiale | Recycled Plastic |
| Forma | Rettangolare |
| Sovrapponibile | Sì |
| N. per custodia | 12 Racks |