missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FLJ13910 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Marca: Abnova™ H00064795-T02.100uL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
Western Blot | |
BC012165 | |
pCMV-FLJ13910 full-length | |
100 μL | |
Human |
FLJ13910 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 43.12kDa | |
Transient overexpression cell lysate was tested with anti-FLJ13910 antibody (Mfr. No. H00064795-B01P) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |