missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FLJ23588 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Marca: Abnova™ H00064800-T01.100uL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
293T | |
FLJ23588 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 155.8kDa | |
Transient overexpression cell lysate was tested with anti-FLJ23588 antibody (Mfr. No. H00064800-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
BC039315 | |
pCMV-FLJ23588 full-length | |
100 μL | |
Human |