Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Marca: Eppendorf™ 6001011110
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), DWP (53 mm), side access, stainless steel- Rack for DWP (53 mm/ 2 in)- Max. DWP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 549 x 139 x 389 (21.6 x 5.5 x 15.3 in)- Max. DWP number per rack: 42 (6 x 7)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 378 DWP), Innova U535 (max. 504 DWP), Innova U725 and U725-G (max. 756 DWP), CryoCube F740 series (3-compartment version; max. 756 DWP)Specifica
Freezer Rack |