Learn More
3M™ Full Face Respirators
Marca: 3M™ 7000108594
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- The popular 3M 6000 series of low maintenance respirators now includes a range of lightweight full face silicone respirators. For workers who are exposed to gases, vapours, fine particles or any combination of these.
- Lightweight
- Cost effective
- Wide field of vision
- Excellent visibility
- Easy adjustment
- Comfortable fit
- Tested in accordance with EN 140
- Easy to use bayonet fitting system
Specifica
Silicone, Thermoplastic Elastomer | |
Fullmask | |
Twin lightweight filters or a supplied air system |