Learn More
Univet Optical Technologies™ Occhiali con lenti G15
Codice prodotto. 12696306
Click to view available options
:
White/Blue
Codice prodotto. 12696306
Marca: Univet Optical Technologies™ 5X7.03.22.00
Protective eyewear designed to be worn over your prescription lenses and featuring maximum protection and comfort. Univet Optical Technologies™5X7 Ultimate OTG Protective Eyewear is adjustable and uses SoftPad technology.
  • The ultimate in over-spectacle protective eyewear
  • Maximum protection and comfort
  • Carefully designed to prevent overlapping of the temples
  • SoftPad technology
  • Adjustable temple length
Specifica
Certificazione/Conformità EN 166, EN 167, EN 168, EN170
Lens Coating Antigraffio
Lens Tint Clear
Product Type Occhiali
Da utilizzare con (applicazione) Uso generico e protezione meccanica
Colore telaio Bianco/Blu
N. per confezione 1 ciasc.
Lens Material Policarbonato