Learn More
GAB1 & SOS1 Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Marca: Abnova DI0185.1Set
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the GAB1 protein, and the other against the SOS1 protein for use in in situ Proximity Ligation Assay.Specifica
GAB1 | |
Unconjugated | |
- | |
1 Set | |
Primary | |
Antibody pair set content: 1. GAB1 rabbit purified polyclonal antibody (20 ug) 2. SOS1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
In situ PLA | |
GRB2-associated binding protein 1 | |
GAB1 | |
RUO | |
2549/6654 | |
Human | |
Ab Pair |