missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-1 Mouse anti-Human, Clone: 3G10D2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60223-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.Specifica
Galectin-1 | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09382 | |
LGALS1 | |
Galectin-1 Fusion Protein Ag2432 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3G10D2 | |
Unconjugated | |
LGALS1 | |
14 kDa laminin binding protein, 14 kDa lectin, Gal 1, GAL1, Galaptin, Galectin 1, Galectin-1, HBL, HLBP14, HPL, Lactose binding lectin 1, LGALS1, S Lac lectin 1 | |
Mouse | |
Protein G | |
RUO | |
3956 | |
-20°C | |
Liquid |