Learn More
Fisherbrand™ Gamma Radiation Safe Storage Box for Four Way Racks
Provides radiation protection shielding for use in laboratories.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Manufactured with hinged lids
- Transparent shielding
- Serves as solution for short-term storage of radioactive waste or radioisotopes
- Constructed of acrylic provides easy fabrication and manufacturability
Specifica
Hinged lid | |
Gamma radiation shielded | |
Acrylic |
17.5 x 15 x 14cm (6.89 x 5.90 x 5.51 in.) | |
Storage Box for four way racks | |
Radioisotopes or radioactive waste |