Learn More
Thermo Scientific™ Manual, Fixed Needle, Gas Tight Syringes for GC Instruments
Codice prodotto. p-4525961 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Volume (sistema metrico):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
Calibro dell'ago:
22 G
22s G
23 G
23s G
25 G
26 G
26s G
Lunghezza (sistema metrico) ago:
50 mm
57 mm
85 mm
Stile del punto dell'ago:
Bevel
Blunt
Cone
Side Hole
Dimensione della confezione:
Pezzo
Codice prodotto. 12826036
Marca: Thermo Scientific™ 365F2441
Choose Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for manual accurate, reproducible GC sample introduction using an affixed needle and a plunger that creates a tight seal.
Get durability, clarity and accuracy from Thermo Scientific™ Manual Fixed Needle, Gas Tight Syringes. Used for manual sample injection into gas chromatography (GC) instrument, they provide accuracy and reproducibility that gives you confidence in analytical results, time-after-time. These syringes have an affixed needle and a plunger that creates a tight seal with the barrel. Choose from a variety of sizes and types. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
Durability
The enhanced design of the syringes delivers:
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
Clarity
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
Accuracy
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover:
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
TRUSTED_SUSTAINABILITY
Specifica
Descrizione 25 μL; 57 mm L; 23s gauge fixed
Calibro dell'ago 23s G
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Quantità Ea.
Volume (sistema metrico) 25 μL
Lunghezza (sistema metrico) ago 57 mm
Stile del punto dell'ago Cone
Da utilizzare con (apparecchiatura) TriPlus RSH Autosampler and TriPlus 100LS Autosampler