Learn More
Biodesign™ Gel storage system GelFile binder only with clear label holder BioDesign
Marca: Biodesign™ G305
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Stores, organises and displays dried polyacrylamide and agarose gels- Each BioDesign GelFile™ binder contains heavy crystal clear vinyl pages with pockets and white backgrounds
- The pages are chemically neutral and are guaranteed not to alter your dried gels or turn yellow for TEN years
- Dried gels are held flat for long term storage without curling
- Shields gels from moisture, accidental spills and possible damage
- White background allows optimal comparison between gels
- Each BioDesign GelFile™ binder holds 20 to 25 pages
- A clear label holder on the spine allows you to personalise each binder
Specifica
Gel Storage Binder | |
1 Binder |