Learn More
Hirschmann™ Matraccio volumetrico graduato in vetro marrone con stopper in polietilene
Codice prodotto. 11912043
Click to view available options
Capacità (sistema metrico):
10 ml
10,000 mL
100 ml
1000 mL
20 mL
200 mL
2000 mL
25 mL
250 mL
5 mL
50 mL
500 ml
5000 mL
Giunto rastremato standard:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
19/32 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
NS 14/23
Smeriglio standard 10/19 NS
Dimensione della confezione:
Pezzo
pezzi 2
Codice prodotto. 11912043
Marca: Hirschmann™ 2822161
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
  • With polyethylene stopper
  • With amber stain graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifica
Certificazione/Conformità DIN EN ISO 1042
Materiale chiusura Polietilene
Capacità (sistema metrico) 10 ml
Materiale Vetro Duran, Duran Glass
Altezza (sistema metrico) 90 mm
Limite di errore ±0,04 ml
Giunto rastremato standard Smeriglio standard 10/19 NS
Tolleranza ±0.04 ml
Graduated
Classe A