missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GMPR 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Marca: Abnova™ H00002766-T01.100uL
Dettagli aggiuntivi : Peso : 0.00010kg
Specifica
Western Blot | |
GMPR 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 38.06kDa | |
Transient overexpression cell lysate was tested with anti-GMPR antibody (Mfr. No. H00002766-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
NM_006877.2 | |
pCMV-GMPR full-length | |
100 μL | |
Human |