Learn More
SOCS1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SOCS1.
Marca: Abnova PAB6012.100ug
Descrizione
This gene encodes a member of the STAT-induced STAT inhibitor (SSI), also known as suppressor of cytokine signaling (SOCS), family. SSI family members are cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by a subset of cytokines, including IL2, IL3 erythropoietin (EPO), CSF2/GM-CSF, and interferon (IFN)-gamma. The protein encoded by this gene functions downstream of cytokine receptors, and takes part in a negative feedback loop to attenuate cytokine signaling. Knockout studies in mice suggested the role of this gene as a modulator of IFN-gamma action, which is required for normal postnatal growth and survival. [provided by RefSeq
Sequence: C-VLRDYLSSFPFQISpecifica
| SOCS1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western blot (1-3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| SOCS1 | |
| SOCS1 | |
| A synthetic peptide corresponding to C terminus of human SOCS1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of SOCS1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| CIS1/CISH1/JAB/SOCS-1/SSI-1/SSI1/TIP3 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8651 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only