missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GPATC3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Marca: Abnova™
Dettagli aggiuntivi : Peso : 0.00010kg
Specifica
Western Blot | |
GPATC3 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 59.3kDa | |
Transient overexpression cell lysate was tested with anti-GPATC3 antibody (Mfr. No. H00063906-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
BC007767 | |
pCMV-GPATC3 full-length | |
100 μL | |
Human |