missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA552497
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: KRKDYLKQHM KTHAPERDVC RCPREGCGRT YTTVFNLQSH ILSFHEESRP FVCEHAGCGK TFAMKQSLTR HAVVHDPDKK KMKLKVKKSR EKRSLASHLS GYIPPKRKQG QGLSLCQNGE SPNCVEDKML STV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.
The product of this gene is a zinc finger protein with nine Cis[2]-His[2] zinc finger domains. It functions as an RNA polymerase III transcription factor to induce transcription of the 5S rRNA genes. The protein binds to a 50 bp internal promoter in the 5S genes called the internal control region (ICR), and nucleates formation of a stable preinitiation complex. This complex recruits the TFIIIC and TFIIIB transcription factors and RNA polymerase III to form the complete transcription complex. The protein is thought to be translated using a non-AUG translation initiation site in mammals based on sequence analysis, protein homology, and the size of the purified protein.Specifica
GTF3A | |
Polyclonal | |
Unconjugated | |
GTF3A | |
AP2; general transcription factor IIIA; GTF3A; TF3A; TFIIIA; Transcription factor IIIA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92664 | |
GTF3A | |
Recombinant protein corresponding to Human GTF3A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only