Learn More
Guardian Equipment Faucet-Mount Personal Eyewash with Gooseneck-mounted Outlets
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Mounting:
Faucet
Dimensione della confezione:
Pezzo
Descrizione
- Water pressure keeps the stream flowing.
Specifica
Specifica
| Certificazione/Conformità | ANSI Z358.1-2004 as Personal Eyewash Only |
| Collegamento ingresso (sistema imperiale) | 55/64-27 in. (F) |
| Materiale | Chrome-plated Brass |
| Mounting | Faucet |
| Product Type | Eyewash Station |