Learn More
Erlab™ H14 HEPA Filter
Erlab™ H14 HEPA Filter can be use with a range of filtrating fume hoods and storage cabinets.
Marca: Erlab™ H11074031
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Filter HP - HEPA H14 : This filtration technology traps particles larger than 0.1 μm with 99.995
- For use with powders
Specifica
GF4/HP | |
Filter | |
Powders |