Learn More
SCHOTT AG Lighting and Imaging™ Halogen Filter for Spotlight
Halogen Filter for use with EasyLED single or double Spotlight Plus. Convert neutral white LED light to the more yellowish halogen type light. Mimics the color temperature of a halogen light. Available in Red, Blue, Green, Yellow or Halogen and a polarization filter.
Marca: SCHOTT AG Lighting and Imaging™ 600690
Dettagli aggiuntivi : Peso : 0.00660kg
Descrizione
- Color filters for EasyLED single or double Spotlight Plus.
- Use for fixing at spot light with thread.
- Available colors: red, blue, green, yellow and halogen
- Polarization filter available.
Specifica
Halogen Filter | |
Convert neutral white LED light to the more yellowish halogen type light | |
EasyLED Spotlight Plus (Single or Double) |
Orange | |
Stereo Microscopy Illumination |