Learn More
Hamilton™ Metal Hub Sharp Non-Coring Needles (Luer Lock)
Codice prodotto. 10571485
Click to view available options
Calibro dell'ago:
19 G
20 G
21 G
22 G
22s G
23 G
24 G
25 G
26 G
26s G
27 G
31 G
Da utilizzare con (apparecchiatura):
LT and TLL Syringes and Connectors
PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors
Autoclavabile:
Autoclavable
Not Autoclavable
Dimensione della confezione:
pezzi 3
pezzi 6
Codice prodotto. 10571485
Marca: Hamilton™ 90039
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. Needles are 304 stainless steel (sst.). Standard needle length is 2 in. (51mm). Custom needle lengths and point styles are available.
Metal Hub Compatible Syringe Terminations
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
  • Needle Point Styles
    • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 33-10
    /li>
    Six needles per pack.
    Specifica
    Autoclavabile Autoclavable
    Materiale Nickel-plated Brass (Hub), Stainless Steel
    Calibro dell'ago 26s G
    Diametro (sistema imperiale) interno 0.005 in.
    Diametro (sistema imperiale) esterno 0.018 in.
    Diametro (sistema metrico) interno 0.13 mm
    Diametro (sistema metrico) esterno 0.45 mm
    Lunghezza (sistema imperiale) ago 2 in.
    Product Type Replacement Needle
    Stile del punto dell'ago Sharp, Bevel, Curved
    Da utilizzare con (apparecchiatura) PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors