Learn More
HAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 25133-1-AP-20UL
 vedi altre versioni di questo prodotto
Codice prodotto. 16883675
  • 145.00€ / 20µL
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Huntington′s disease (HD), a neurodegenerative disorder characterized by loss of striatal neurons, is caused by an expansion of a polyglutamine tract in the HD protein huntingtin. HAP1 was initially identified through a two-hybrid library screening; the binding of HAP1 to huntingtin correlated with the expansion of the polyglutamine tract. HAP1 also interacts with two cytoskeletal proteins (dynactin and pericentriolar autoantigen protein 1), suggesting that HAP1 may play a role in vesicular trafficking or organelle transport. HAP1 is also involved with the huntingtin-enhanced BDNF transport along the cellular microtubles. Attenuation of this process led to the loss of neurotrophic support and neuronal toxicity, which suggests that loss of this function might contribute to pathogenesis. Several alternatively spliced isoforms have been described for HAP1.
Specifica
Specifica
HAP1
Polyclonal
Unconjugated
HAP1
HAP 1, HAP1, HAP2, hHLP1, HIP5, HLP, HLP1, Neuroan 1
Rabbit
Antigen Affinity Chromatography
RUO
15114, 9001
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35668, P54257
HAP1
HAP1 Fusion Protein Ag17705
20 μL
Primary
Mouse, Human
Antibody
IgG