missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HAPLN3 Recombinant Protein

Marca:  Abnova™ H00145864-P01-25

 vedi altre versioni di questo prodotto

Codice prodotto. 16128403

  • 521.00€ / 25µg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Esplora altre offerte speciali
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH62320.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
63.47
8
Glutathione Sepharose 4 Fast Flow
25 μg
LPFYNGFYYSNSANDQNLGNGHGKDLLNGVKLVVETPEETLFTYQGASVILPCRYRYEPALVSPRRVRVKWWKLSENGAPEKDVLVAIGLRHRSFGDYQGRVHLRQDKEHDVSLEIQDLRLEDYGRYRCEVIDGLEDESGLVELELRGVVFPYQSPNGRYQFNFHEGQQVCAEQAAVVASFEQLFRAWEEGLDWCNAGWLQDATVQYPIMLPRQPCGGPGLAPGVRSYGPRHRRLHRYDVFCFATALKGRVYYLEHPEKLTLTEAREACQEDDATIAKVGQLFAAWKFHGLDRCDAGWLADGSVRYPVVHPHPNCGPPEPGVRSFGFPDPQSRLYGVYCYRQH
EXLD1/HsT19883
HAPLN3
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
145864
HAPLN3 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HAPLN3
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.