missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HARS (Human) Recombinant Protein

Human HARS full-length ORF ( AAH11807, 1 a.a. - 509 a.a.) recombinant protein with GST-tag at N-terminal.

344.00€ - 521.00€

Specifications

Numero di accesso AAH11807
ID gene (immissione) 3035
Nome histidyl-tRNA synthetase
Metodo di preparazione Wheat germ expression system
Analisi di controllo qualità 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantità Price Quantity & Availability  
Product Code Brand Quantità Price Quantity & Availability  
16116001
View Documents
Abnova™
H00003035-P01.10ug
10 μg
344.00€
10µg
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16126001
View Documents
Abnova™
H00003035-P01.25ug
25 μg
521.00€
25µg
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

  • Sequence: MAERAALEELVKLQGERVRGLKQQKASAELIEEEVAKLLKLKAQLGPDESKQKFVLKTPKGTRDYSPRQMAVREKVFDVIIRCFKRHGAEVIDTPVFELKETLMGKYGEDSKLIYDLKDQGGELLSLRYDLTVPFARYLAMNKLTNIKRYHIAKVYRRDNPAMTRGRYREFYQCDFDIAGNFDPMIPDAECLKIMCEILSSLQIGDFLVKVNDRRILDGMFAICGVSDSKFRTICSSVDKLDKVSWEEVKNEMVGEKGLAPEVADRIGDYVQQHGGVSLVEQLLQDPKLSQNKQALEGLGDLKLLFEYLTLFGIDDKISFDLSLARGLDYYTGVIYEAVLLQTPAQAGEEPLGVGSVAAGGRYDGLVGMFDPKGRKVPCVGLSIGVERIFSIVEQRLEALEEKIRTTETQVLVASAQKKLLEERLKLVSELWDAGIKAELLYKKNPKLLNQLQYCEEAGIPLVAIIGEQELKDGVIKLRSVTSREEVDVRREDLVEEIKRRTGQPLCIC
Specifications

Specifications

AAH11807
histidyl-tRNA synthetase
125% SDS-PAGE Stained with Coomassie Blue
FLJ20491, HRS
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
3035
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HARS
GST
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.