Learn More
Taylor Wharton™ HC35 High Capacity Cryostorage System
Ruggedly constructed, high-capacity cryogenic storage dewar. Taylor Wharton™ HC35 High Capacity Cryostorage System uses a convenient canister index location ring and a tapered internal spider.
Marca:  Taylor Wharton™ HC35
Codice prodotto. 11728519
  • 2940.00€ / Pezzo
Spedizione stimata: 30-04-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Large storage capacity.
  • New Rugged Construction - ribbed high strength aluminum body, magniformed necktube design, and more durable paint.
  • Versatile Storage System - convenient canister index location ring and tapered internal spider.
  • Superior vacuum performance with super insulation provides maximum holding times.
  • Security - Optional low-level alarm is available with remote monitor capability.
  • Mobility - Roller bases are available for all models
Specifica
Specifica
35 L
681 mm
17.7/46 kg (empty/full)
Vials (1020 x 2 mL)
10 canisters, 67 x 279mm
478 mm
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Vedi i prodotti accessori