missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hCG beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA558593
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: PTMTRVLQGV LPALPQVVCN YRDVRFESIR LP Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 59%.
hCG beta is the beta chain of hCG (Human chorionic gonadotropin), a 40 kDa glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. hCG is made up of alpha and beta subunits that are non-covalently associated. The alpha subunit is similar to those found on the pituitary hormones FSH, LH ans TSH. However, the beta subunit is different in FSH, LH and TSH and are involved in conferring biological specificity. Maintece of the fetus in the first trimester of pregcy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium. hCG is present only in trace amounts in non-pregt urine and sera but rises sharply during pregcy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). Diseases associated with Hcg dysfunction include gestational trophoblastic tumor and hydatidiform mole, recurrent 1.Specifica
hCG beta | |
Polyclonal | |
Unconjugated | |
CGB3 | |
Chorionic Gonadotropin, Beta Polypeptide, Choriogonadotropin Subunit Beta Variant, FSH-alpha, LSH-alpha, TSH-alpha, anterior pituitary glycoprotein hormones common subunit alpha, choriogonadotropin alpha chain, chorionic gonadotrophin subunit alpha, chorionic gonadotropin, alpha polypeptide, follicle-stimulating hormone alpha chain, follicle-stimulating hormone alpha subunit, follitropin alpha chain, glycoprotein hormones alpha chain, luteinizing hormone alpha chain, lutropin alpha chain, thyroid-stimulating hormone alpha chain, thyrotropin alpha chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0DN86 | |
CGB3 | |
Recombit protein corresponding to Human hCG beta | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto