Learn More
Abnova™ HDAC8 Recombinant Protein
Recombinant protein for HDAC8 (human) gene
344.00€ - 521.00€
Specifica
Numero di accesso | AAH50433 |
---|---|
Da utilizzare con (applicazione) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulazione | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
ID gene (immissione) | 55869 |
Peso molecolare | 66.99 |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16171913
|
Abnova™
H00055869-P01.10ug |
10 μg |
344.00€
10µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
16181913
|
Abnova™
H00055869-P01.25ug |
25 μg |
521.00€
25µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA.
- Human HDAC8 full-length ORF ( AAH50433, 1 a.a. - 377 a.a.) recombinant protein with GST-tag at N-terminal
- histone deacetylase 8
- Theoretical molecular weight: 66.99kDa
- Preparation: in vitrowheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer
Sequence: MEEPEEPADSGQSLVPVYIYSPEYVSMCDSPAKIPKRASMVHSLIEAYALHKQMRIVKPKVASMEEMATFHTDAYLQHLQKVSQEGDDDHPDSIEYGLGYDCPATEGIFDYAAAIGGATITAAQCLIDGMCKVAINWSGGWHHAKKDEASGFCYLNDAVLGILRLRRKFERILYVDLDLHHGDG VEDAFSFTSKVMTVSLHKFSPGFFPGTGDVSDVGLGKGRYYSVNVPIQDGIQDEKYYQICESVLKEVYQAFNPKAVVLQLGADTIAGDPMCSFNMTPVGIGKCLKYILQWQLATLILGGGGYNLANTARCWTYLTGVILGKTLSSEIPDHEFFTAYGPDYVLEITPSCRPDRNEPHRIQQILNYIKGNLKHVV
Best use within three months from the date of receipt.
ELISA, western blotting (recombinant protein), antibody production, protein array
Specifica
AAH50433 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
66.99 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HDAC8 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
55869 | |
HDAC8 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEEPEEPADSGQSLVPVYIYSPEYVSMCDSPAKIPKRASMVHSLIEAYALHKQMRIVKPKVASMEEMATFHTDAYLQHLQKVSQEGDDDHPDSIEYGLGYDCPATEGIFDYAAAIGGATITAAQCLIDGMCKVAINWSGGWHHAKKDEASGFCYLNDAVLGILRLRRKFERILYVDLDLHHGDGVEDAFSFTSKVMTVSLHKFSPGFFPGTGDVSDVGLGKGRYYSVNVPIQDGIQDEKYYQICESVLKEVYQAFNPKAVVLQLGADTIAGDPMCSFNMTPVGIGKCLKYILQWQLATLILGGGGYNLANTARCWTYLTGVILGKTLSSEIPDHEFFTAYGPDYVLEITPSCRPDRNEPHRIQQILNYIKGNLKHVV | |
HDACL1/RPD3 | |
HDAC8 | |
Wheat Germ (in vitro) | |
GST |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.