missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Heated Product Shelf Chambers
Allows drying of bulk samples, or drying of serum bottles, vials or microplates. Labconco Heated Product Shelf Chambers are wireless and removable so that they may be placed in freezer for pre-freezing.
Marca: Labconco™ 780801130
Includes: 5 shelves and clear chamber
Requires : FreeZone Freeze Dryer
Descrizione
- Short Chambers include three 10 in. diameter shelves and three temperature probes; Tall Chambers include five 10 in. diameter shelves and five temperature probes
- Shelves are wireless and removable so that they may be placed in a freezer for pre-freezing
- Each shelf is height adjustable to accommodate broad variety of sample sizes
- Chamber connects to FreeZone Freeze Dryer for control and recording of shelf temperature from Lyo-Works display
- Temperature may be set from −20° C to 60° C (−4° F to 140° F)
- Each Chamber includes 12 in. diameter clear acrylic cylinder, vacuum release valve and 8 in. diameter opening to collector on bottom of chamber
- Will fit 10.12 in. or 14.0 in. diameter condensers
- For use with any FreeZone Freeze Dryer except Legacy models
Compliance
All models conform to UL Standard 61010-1 (60 Hz models), CAN/CSA C22.2 No. 61010-1 (60 Hz models), and CE Conformity marking (230V, 50 Hz models).
Servizi
One year
Specifica
Heated Shelf | |
-20°C to +60°C | |
UL, CAN/CSA, CE | |
15.2 in. | |
15.2 in. | |
24.6 in. | |
2A | |
230 V | |
80 lb. |
3 | |
Steel (Shelves), Acrylic (Chamber) | |
5 shelves and clear chamber | |
38.6 cm | |
38.6 cm | |
62.4 cm | |
Schuko | |
50/60 Hz | |
36.29 kg |