Learn More
Heidolph™ Instruments Hei-VAP Ultimate Control ML G3, EU-plug
The Hei-VAP Ultimate Control ML G3 adds numerous smart features to the Hei-VAP Ultimate.
Marca:  Heidolph™ Instruments 576-02300-00
Dettagli aggiuntivi : Peso : 19.72000kg
Codice prodotto. 17343370
  • 6180.00€ / Pezzo
Spedizione stimata: 29-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Adds numerous smart features to the Hei-VAP Ultimate, as well as an overview of all parameters and the current process.
  • 7 in. touch display for controlling the entire system including the vacuum and the cooling temperature
  • Motor lift model with G3 vertical glassware
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start screen
  • Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
  • Define and save own evaporation processes (ramps)
  • Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
  • USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
Specifica
Specifica
5 L
Motor Lift Rotary Evaporator
EU-plug