Learn More
Thermo Scientific™ Herasafe™ KS Options and Accessories
Codice prodotto. 10627824 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Product Type:
1200 mm Indented Worksurface Module
150 mm Flat Worksurface Module
1500 mm Indented Worksurface Module
1800 mm Indented Worksurface Module
300 mm Flat Worksurface Module
300 mm Vubration Damped Worksurface
600 mm Flat Worksurface Module
600 mm Indented Worksurface Module
600 mm Vubration Damped Worksurface
900 mm Flat Worksurface Module
900 mm Indented Worksurface Module
Utility Basket
Utility Basket with Hanging Bar
Dimensione della confezione:
Pezzo
Codice prodotto. 10627824
Marca: Thermo Scientific™ 50073666
Worksurfaces, exhaust transitions and baskets
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Flat Worksurface Module 150 mm Wide (316SS)
Product Type 150 mm Flat Worksurface Module
Larghezza (sistema metrico) parte esterna 150 mm
Materiale alloggiamento 316 SS
Larghezza (sistema metrico) 150 mm