Learn More
Thermo Scientific™ Herasafe™ KS Options and Accessories
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Product Type:
1200 mm Indented Worksurface Module
150 mm Flat Worksurface Module
1500 mm Indented Worksurface Module
1800 mm Indented Worksurface Module
300 mm Flat Worksurface Module
300 mm Vubration Damped Worksurface
600 mm Flat Worksurface Module
600 mm Indented Worksurface Module
600 mm Vubration Damped Worksurface
900 mm Flat Worksurface Module
900 mm Indented Worksurface Module
Utility Basket
Utility Basket with Hanging Bar
Dimensione della confezione:
Pezzo
Descrizione
Worksurfaces, exhaust transitions and baskets
Specifica
Specifica
| Descrizione | Indented Worksurface Module 600 mm Wide (316SS) |
| Product Type | 600 mm Indented Worksurface Module |
| Larghezza (sistema metrico) parte esterna | 600 mm |
| Materiale alloggiamento | 316 SS |
| Larghezza (sistema metrico) | 600 mm |