missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taylor Wharton™ High Capacity Refrigerator
Marca: Taylor Wharton™ VHC35
Dettagli aggiuntivi : Peso : 0.80000kg
Specifica
Stores up to (1,260) 1.2ml vials (9,900) 1/2 cc straws | |
VHC 35 | |
26.8 in. |
130 days | |
35 L |