Learn More
Thermo Scientific Chemicals High Form Nickel Crucible;Cap (ml), 50;Top Dia (mm), 44;Bottom Dia (mm), 32;Depth (mm), 51
Click to view available options
Capacità (sistema metrico):
20 mL
30 mL
50 mL
75 mL
100 mL
250 mL
500 mL
Diametro (sistema metrico) esterno:
38mm
41mm
44mm
54mm
61mm
84mm
101mm
Altezza (sistema metrico):
35mm
43mm
51mm
57mm
62mm
78mm
91mm
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Descrizione | High Form |
| Materiale | Nickel |
| Capacità (sistema metrico) | 50 mL |
| Diametro (sistema metrico) esterno | 44mm |
| Altezza (sistema metrico) | 51mm |
| Product Type | Nickel Crucible |
| Diametro (sistema metrico) base | 32mm |