Learn More
Epredia™ Histoplast Paraffin
Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.
Brand:  Epredia™ 8331
 View more versions of this product
Product Code. 12693026
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Made of paraffin and plastic polymers
  • Delivers excellent processing and yields thin, wrinkle-free sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).
Specifications
Specifications
56°C
Paraffin
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers