missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HL-60 (human promyelocytic leukemia) Whole cell lysate, Non-denatured; Abnova
Non-denatured whole cell lysate of HL-60 (human promyelocytic leukemia)
Marca: Abnova L014V2.200ug
This item is not returnable.
View return policy
Description
- Tissue: Peripheral Blood
- Host: Human
- Lysis Buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA lysis buffer
Western Blotting
Specifications
Western Blot | |
Whole cell lysate (non-denatured) | |
Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). | |
200 μg | |
Human |
Blood, peripheral | |
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
In modified RIPA Lysis Buffer. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction