Learn More
OHAUS™ Holder Clamp
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Materiale:
Aluminum
Nickel-plated
Stainless Steel
Stile:
All Position
Jumbo
Regular
Shelf Grip
Swivel
Ultra Clamp
Universal
Dimensione della confezione:
Pezzo
Descrizione
- Stainless steel is autoclavable and corrosion-resistant
- Aluminum is lightweight yet strong
- Cast iron with black enamel finish is chemical-resistant
- Available in different shapes and sizes to fit your laboratory needs
Specifica
Specifica
| Stile | Universal |
| Tipo | Holder |
| Materiale | Aluminum |
| Descrizione | Clamp, Holder, Universal, CLC-UNSMBA |
| Size | Universal |
| Dimensioni (sistema metrico) impugnatura | 0 to 22 mm |