Learn More
Sperian™ Schermo facciale Honeywell™ Clearways
10.10 EUR valido fino al 2024-07-31
Usa il codice promo "21640" per avere il tuo prezzo promozionale.
Usa il codice promo "21640" per avere il tuo prezzo promozionale.
Click to view available options
Product Type:
Schermo facciale Clearways
Lens Coating:
Anti-fog
Uncoated
Lens Material:
Acetate
N/A
Polycarbonate
Descrizione
- Recommended for the agriculture, chemical and forestry industries
- Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
- Safe, practical, economical integral visor fitting ensures extra protection
Order Info
Note: The image shows both the Browguard and the Visor. Both pieces must be ordered separately. See Browguard (11886980), and Visors (11806990, 12652733, or 11896980).
Specifica
Certificazione/Conformità | EN 166 1 F |
Lens Coating | Anti-fog |
Product Type | Schermo facciale Clearways |
Modello | CV84AN |
N. per confezione | Confezione da 1 |
Lens Material | Acetate |