Learn More
Honeywell™ Schermo facciale Honeywell™ Supervizor
Click to view available options
Modello:
SV9AC
SV9PC
Descrizione:
200 mm Clear Acetate Visor
230 mm Clear Polycarbonate Visor
Browguard with Ratchet Headband
Lens Material:
Acetate
Policarbonato
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
- Recommended for use in the chemical and forestry industries, among others
- Protects against hazards such as abrasion, airborne particles, biohazards, chemicals, gases, vapors, and smoke
- Maximum protection, allowing full unrestricted head movement
- Creased visor eliminates gaps and visor profile follows contours of the face
Specifica
Specifica
| Certificazione/Conformità | EN 166 1B, EC 0100, EC 0086 |
| Larghezza (sistema metrico) | 230 mm |
| Descrizione | 230 mm Clear Polycarbonate Visor |
| Product Type | Schermo facciale Supervizor |
| Modello | SV9PC |
| N. per confezione | 1 ciasc. |
| Lens Material | Policarbonato |