Learn More
Asecos™ Housing Unit Adapter for Recirculaing Air Filter Systems
Adapter as a separate housing unit for lateral mounting, suitable for cabinets with a depth of 574 mm.
Marca: Asecos™ 26885
583.80 EUR valido fino al 2024-09-17
Usa il codice promo "21690" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Accessory item
- Housing made of powder coated sheet steel
Specifica
Housing unit | |
Lateral mounting | |
Steel | |
Recirculating air filter systems |