Learn More
Schmidlin™ HP Plus Tower Nitrogen Generator
This nitrogen generator produces a continuous supply of high purity nitrogen.
Marca:  Schmidlin™ DB-N2T-750-EU
Dettagli aggiuntivi : Peso : 15.00000kg
Codice prodotto. 17778899
  • 8482.06€ / Pezzo
Spedizione stimata: 14-10-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Purity: up to +99.999%
  • Pressure: up to 5 barg (75 psig)
  • Ideal for all GC applications
  • Stable baseline with increased sensitivity and repeatability
  • Meets and exceeds requirements for the most demanding GC applications
Specifica
Specifica
63 cm
49 cm
5 bar
140 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
12 W
750 mL/min.
35°C
55 in.
Inlet: 1/4 in., Outlet: 1/8 in.
>99.999%