Learn More
Rotronic™ HP23-AW Set 40 Thermo Hygrometer Set
Perfect solution for on-site measurements, this hygrometer is supplied in a tough, lightweight ABS carrying case. Rotronic™ HP23-AW Set 40 Thermo Hygrometer Set includes everything needed for measurement and calibration.
Marca: Rotronic™ HP23-AW-SET-40FS
Descrizione
- Handheld instrument
- Measurement probe
- Sample holder
- Disposable sample container
- Humidity standards (10%rh, 35%rh, 50%rh, 80%rh)
- Carrying case
Specifica
Hygrometer Set | |
ABS Housing | |
5 m | |
Handheld instrument, probe, sample holder, sample container, humidity standards, carrying case |
9V Alkaline | |
270 x 70 x 30 mm | |
198 g |