Learn More
SCAT Starter Kits HPLC - Fornitura, V2.0
Sfoglia Tutto SCAT Europe Prodotti
Click to view available options
Tipo:
Solvent Waste Kit
Starter Kit HPLC 1
Starter Kit HPLC 2
Starter Kit HPLC 3
Dimensione della confezione:
1 set
pezzi 4
Descrizione
- Original brand quality from the premium manufacturer SCAT
- Initial removal equipment that can be used with all common HPLC system
- Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
- All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifica
Specifica
| Include | HPLC Starter Kit 4: 4 x Safety Cap IV, GL45, 4 x air valve, 4 x change indicator, 16 x PFA fitting 1.6 mm OD, green, 16 x PFA fitting 2.3 mm OD, violet, 16 x PFA fitting 3.2 mm OD, blue, 12 x PFA blind plugs, 4 x labelling field |
| Materiale | PTFE |
| Dimensioni filo | GL45 |
| Tipo | Solvent Waste Kit |
| Quantità | 4 Pk. |