Learn More
Abnova™ HTR1D (Human) Recombinant Protein
Human HTR1D full-length ORF ( AAH07720.1, 1 a.a. - 377 a.a.) recombinant protein with GST-tag at N-terminal.
Marca:  Abnova™ H00003352-P01.10ug
Codice prodotto. 16126481
  • 335.00€ / 10µg
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Sequence: MSPLNQSAEGLPQEASNRSLNATETSEAWDPRTLQALKISLAVVLSVITLATVLSNAFVLTTILLTRKLHTPANYLIGSLATTDLLVSILVMPISIAYTITHTWNFGQILCDIWLSSDITCCTASILHLCVIALDRYWAITDALEYSKRRTAGHAATMIAIVWAISICISIPPLFWRQAKAQEEMPDCLVNTSQISYTIYSTCGAFYIPSVLLIILYGRIYRAARNRILNPPSLYGKRFTTAHLITGSAGSSLCSLNSSLHEGHSHSAGSPLFFNHVKIKLADSALERERISAARERKATKILGIILGAFIICWLPFFVVSLVLPICRDSCWIHPALFDFFTWLGYLNSLINPIIYTVFNEEFRQAFQKIVPFRKAS
Specifica
Specifica
AAH07720.1
5-hydroxytryptamine (serotonin) receptor 1D
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HTR1D
GST
3352
Wheat germ expression system
10 μg
5-HT1D, HT1DA, HTR1DA, HTRL, RDC4
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer