missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DPYSL5 Full-length ORF (AAH02874, 1 a.a. - 564 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00056896-P01.25ug

 vedi altre versioni di questo prodotto

Codice prodotto. 16172303

  • 508.00€ / 25µg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

Members of the CRMP family, such as DPYSL5, are believed to play a role in growth cone guidance during neural development.[supplied by OMIM]

Sequence: MLANSASVRILIKGDKVVNDDCTHEADVYIENGIIQQVGRELMIPGGAKVIDATGKLVIPGGIDTSTHFHQTFMNATCVDDFYHGTKAALVGGTTMIIGHVLPDKETSLVDAYEKCRGLADPKVCCDYALHVGITWWAPKVKAEMETLVREKGVNSFQMFMTYKDLYMLRDSELYQVLHACKDIGAIARVHAENGELVAEGAKEALDLGITGPEGIEISRPEELEAEATHRVITIANRTHCPIYLVNVSSISAGDVIAAAKMQGKVVLAETTTAHATLTGLHYYHQDWSHAAAYVTVPPLRLDTNTSTYLMSLLANDTLNIVASDHRPFTTKQKAMGKEDFTKIPHGVSGVQDRMSVIWERGVVGGKMDENRFVAVTSSNAAKLLNLYPRKGRIIPGADADVVVWDPEATKTISASTQVQGGDFNLYENMRCHGVPLVTISRGRVVYENGVFMCAEGTGKFCPLRSFPDTVYKKLVQREKTLKVRGVDRTPYLGDVAVVVHPGKKEMGTPLADTPTRPVTRHGGMRDLHESSFSLSGSQIDDHVPERASARILAPPGGRSSGIW
Specifica

Specifica

AAH02874
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.78kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRAM/CRMP-5/CRMP5/FLJ45383/Ulip6
DPYSL5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56896
DPYSL5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLANSASVRILIKGDKVVNDDCTHEADVYIENGIIQQVGRELMIPGGAKVIDATGKLVIPGGIDTSTHFHQTFMNATCVDDFYHGTKAALVGGTTMIIGHVLPDKETSLVDAYEKCRGLADPKVCCDYALHVGITWWAPKVKAEMETLVREKGVNSFQMFMTYKDLYMLRDSELYQVLHACKDIGAIARVHAENGELVAEGAKEALDLGITGPEGIEISRPEELEAEATHRVITIANRTHCPIYLVNVSSISAGDVIAAAKMQGKVVLAETTTAHATLTGLHYYHQDWSHAAAYVTVPPLRLDTNTSTYLMSLLANDTLNIVASDHRPFTTKQKAMGKEDFTKIPHGVSGVQDRMSVIWERGVVGGKMDENRFVAVTSSNAAKLLNLYPRKGRIIPGADADVVVWDPEATKTISASTQVQGGDFNLYENMRCHGVPLVTISRGRVVYENGVFMCAEGTGKFCPLRSFPDTVYKKLVQREKTLKVRGVDRTPYLGDVAVVVHPGKKEMGTPLADTPTRPVTRHGGMRDLHESSFSLSGSQIDDHVPERASARILAPPGGRSSGIW
RUO
DPYSL5
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali