Learn More
Novus Biologicals™ Human Esophageal Tissue MicroArray (Cancer)
Contains esophagus adenosquamous cell carcinoma, moderately differentiated T3N0M0, Stage IIB
Marca: Novus Biologicals™ NBP2-30266
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Formalin fixed
- Species: Human
- Cross reactivity: Human
- Tissue condition: Cancer
- No. of samples: 60
- No. of patients: 60
- Core diameter: 2.0mm
- Section thickness: 4μm
Recommended Storage
Store at 4°C. Do not freeze.
Servizi
1 Year
Specifica
1 Slide | |
Human | |
Human |
RUO | |
Cancer |
Titolo del prodotto
This product is for research use only and is not approved for use in humans or in clinical diagnosis.