missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPRC5D Antibody, R&D Systems™
Mouse Monoclonal Antibody
280.00€ - 678.00€
Specifica
| Antigene | GPRC5D |
|---|---|
| Clone | 571961R |
| Diluizione | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applicazioni | Flow Cytometry, CyTOF |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18713627
|
R&D Systems
MAB6300R-SP |
25 μg |
280.00€
25µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18742298
|
R&D Systems
MAB6300R-100 |
100 μg |
678.00€
100µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Beschreibung
GPRC5D Monoclonal specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Spezifikation
| GPRC5D | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor | |
| NS0 mouse myeloma cell line transfected with human GPRC5D, Met1-Val359, Accession # Q9NZD1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 571961R | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NZD1 | |
| 55507 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts