missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-17A ProQuantum Immunoassay Kit
GREENER_CHOICE

Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument

Marca:  Invitrogen™ A356115

 vedi altre versioni di questo prodotto

Codice prodotto. 17114709

  • 3350.00€ / Pezzo
Spedizione stimata: 12-12-2024
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

  • High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
  • Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
  • Fast, easy workflow: No wash steps, 2 hours from sample to answer
  • Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
  • No proprietary instrument to purchase: Runs on any real-time PCR instrument
  • ProQuantum software available: Robust data analysis and statistical group wise comparison
TRUSTED_SUSTAINABILITY
Specifica

Specifica

Q16552
<0.3 pg/mL
Proximity-based Immunoassay
Human
qPCR Instruments
1 hr.
FAM™
RUO
Frozen at -20°C
30 min.
0.32 to 25,000 pg/mL
Taqman™ Real-time PCR
ProQuantum
Protein quantitation
3605
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard
5 x 96 Tests
Serum,2-5μL; Plasma,2-5μL; Supernatant,2-5μL
Human
2 hr.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
FDS
Documents

Documents

Certificats
Promotions

Promotions