Learn More
Novus Biologicals™ Human Liver Tissue MicroArray (Tumor)
The panel contains 5 to 8 different tissues on one slide.
Marca: Novus Biologicals™ NBP2-78091
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
5 Slides | |
Human | |
Please bake slides at 60C for 30 minutes before use. |